Distillation Apparatus, [ST] 10/30, Ace Glass Incorporated
Supplier: Ace Glass
9322-10 9322-05
89074-392EA 1198.55 CAD
89074-392 89074-440
Distillation Apparatus, [ST] 10/30, Ace Glass Incorporated
Distillation Components Distillation Sets
This apparatus comes has a [ST] 10/30 joint for a 25 mm immersion thermometer
All other joints are [ST] 14/20. The hose connections project to the rear. The round-bottom and pear-shape flasks supplied with the complete unit are 25 mL capacity. The hose connections are for use with 5/16-inch I.D. tubing. Fabricated from borosilicate glass.
Ordering information: Complete apparatus comes complete with 89074-440 distilling head; 89074-590 round bottom flask; and 89074-898 pear shaped flask. Individual components may also be purchased separately.
Order Now
SPECIFICATIONS
Environmentally Preferable