EDTA Standard Solution, 0.0200 N, 4 L, Hach
Supplier: Hach
20556 20556
CA97026-712EA 181.31 CAD
97026-712 CA97026-712
EDTA Standard Solution, 0.0200 N, 4 L, Hach
Standards Environmental Analysis Standards
For hardness determination by buret titration
Hach methods 8222 (calcium) and 8226 (total). 0.0200N (0.0100 M) (N/50).