1-(2-Pyridylazo)-2-naphthol (PAN) solution 0.1% indicator

Supplier: Hach
Danger

2122426 2122449 2122432
CA97026-624EA 73.63 CAD
CA97026-624 CA21224-49 CA97026-644
1-(2-Pyridylazo)-2-naphthol (PAN) solution 0.1% indicator
1-(2-Pyridylazo)-2-naphthol

For determination of low level manganese by the PAN method


Hach method 8149. Range: 0.006 to 0.700 mg/L Mn.

Formula: C₁₅H₁₁N₃O
Storage Temperature: Ambient
MDL Number: MFCD00004071
CAS Number: 85-85-8
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Method 8149
Method Name PAN
Parameter Manganese
Platform Solution
Range 0.006 - 0.700 mg/L Mn

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR