SulfaVer® 4 Sulfate Reagent Powder Pillows, Hach
Supplier: Hach
1206599 2106769
CA12065-99PK 111.39 CAD
CA12065-99 CA21067-69
SulfaVer® 4 Sulfate Reagent Powder Pillows, Hach
Environmental Test Kits
For determination of sulfate by the SulfaVer® 4 turbidimetric method
USEPA accepted for reporting wastewater analysis using Hach Method 8051. Method: SulfaVer® 4. Range: 2 to 70 mg/L SO4.