Copper Reagent Set, Porphyrin, 10 ml, Hach
Supplier: Hach
2603300
CA71002-456EA 857.99 CAD
CA71002-456
Copper Reagent Set, Porphyrin, 10 ml, Hach
Environmental Test Kits
For Copper determination by the Porphyrin Method
Hach Method 8143. Range: 2 - 210 μg/L Cu.
Ordering information: Reagent set includes Copper Masking Reagent Powder Pillows, Porphyrin 1 Reagent Powder Pillows and Porphyrin 2 Reagent Powder Pillows for approximately 100 tests.
Hazardous Items:
Items with this mark may be considered hazardous under some shipping conditions.
If necessary, we will change your selected shipping method to accomodate these items.