Alkaline Cyanide Reagent, Hach
Supplier: Hach
2122326 2122332
CA2122326EA 79.51 CAD
CA2122326 CA2122332
Alkaline Cyanide Reagent, Hach
Environmental Test Kits
For determination of low level manganese by the PAN method
Hach method 8149. Range: 0.006 to 0.700 mg/L Mn.
Ordering information: Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accommodate these items.