Sherlock Bones: Identification of Skeletal Remains Kit
Supplier: BONE CLONES HOLDINGS
COMP-102 COMP-109
470030-032EA 972.8 CAD
470030-032 470030-034 470190-646
Sherlock Bones: Identification of Skeletal Remains Kit
Educational Classroom Kits and Activities
Is the victim a male or female?
  • NGSS Life Science DCIs LS1, and LS2.
  • Pre- and post-laboratory assessments
  • Links to a real world scenario
  • Estimated class/lab time required: 45-60 minutes
This lab provides students with experience with the process of epiphyseal union as well as using Vernier calipers. Students will be given a few bones, and then take the role of a forensic anthropologist to determine as much information as possible about the bones to help identify the individual. Students will become familiar with some of the techniques performed by aMore Product Information