60 Memory Stopwatch, Sper Scientific
Supplier: Sper Scientific
810033 810033C
62370-047EA 197.91 CAD
62370-047 62370-048
60 Memory Stopwatch, Sper Scientific
Timers Digital Stopwatches
Water resistant digital stopwatch features two ¹/₁₀₀th second displays.
  • Counts up or down to 10 hours
  • Ideal for multiple and simultaneous processes
  • Water resistant
In stopwatch mode current lap time shown in upper display, cumulative total in lower one. Memory stores last 60 lap times so there is no need to look away from a test to make notes. It shows fastest, slowest, and average lap times, and has an alarm clock function. In timer mode upper display shows original setting while lower one counts down. Countdown cycles repeat upon completion. 30 second shuttle timer for use during timeouts.
Lap counter shows number of laps (up to 99) in stopwatch mode and number of cycles repeated in timer mode. Pacesetter function can be set to beep continually from 10 beeps per minute to 320 beeps per minute. Shock resistant, superb hand grip.
Certifications: N.I.S.T. traceable certificate of compliance for 62370-048.
Ordering information: Five year warranty.