Silicone Safety Pipette Filler, Sper Scientific
Supplier: Sper Scientific
470113 470114
97035-724EA 74.67 CAD
97035-724 97035-726
Silicone Safety Pipette Filler, Sper Scientific
Pipettes Pipette Bulbs
Pipette fillers enable filling and dispensing of pipettes with one hand.
  • Adjusts to fit any size pipet
  • Chemically resistant glass valves
  • Available in two colors for identification purposes
Safety is ensured through avoidance of hand or mouth contact with liquid. Our Silicone Pipette Fillers offer a softer feel with easier and faster operation of valves. Silicone is also more chemically resistant and more resilient than either PVC or natural rubber. It will not crack or become brittle and will last longer and stay flexible.
Ordering information: Glass tube not included, order separately.