Distillation Receivers, Graduated, Ace Glass
Supplier: Ace Glass
6638-04 6638-06 6638-08
89055-738EA 1888.1 CAD
89055-738 89084-414 89084-416
Distillation Receivers, Graduated, Ace Glass
Distillation Components Distillation Receivers
This is an excellent all-purpose receiver for general distillation work
Body is graduated. Equipped with a complete stopcock manifold to allow contents of the receiver to be removed to a single receiving flask without disturbing the vacuum of the system. Stopcocks are 4mm bore. Tubing connections are 10mm O.D. olive type. Fabricated from borosilicate glass.