DPD Total Chlorine Reagent Powder Pillows, 5 ml, Hach
Supplier: Hach
2198246 1407628
CA21982-46PK 36.07 CAD
CA21982-46 CA97026-702
DPD Total Chlorine Reagent Powder Pillows, 5 ml, Hach
Environmental Test Kits
For determination of total chlorine by the DPD method.
For use with Hach's Chlorine Visual Test Kits.
Range: up to 3.5 mg/L Cl₂.
Caution: *This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.