Liquid-Liquid Extractor Tube, Ace Glass
Supplier: Ace Glass
9414-08 9422-05 9428-12 9451-06
89074-478EA 410.68 CAD
89074-478 89074-488 89074-490 89074-578
Liquid-Liquid Extractor Tube, Ace Glass
Extractors Liquid-Liquid Extractors
This apparatus is used to extract liquids of a ligher density (such as ether) from liquids of a heavier density (such as water)
Fabricated from borosilicate glass.
Ordering information: Complete apparatus consists of: 89074-578 flask (100 mL), 89074-488 extraction chamber, and 89074-478 distilling tube. Individual components may be purchased separately.
Order Now
SPECIFICATIONS
Environmentally Preferable