Sex Pheromone Inhibitor iPD1 Trifluoroacetate
Supplier: Bachem
4018007.0005 4018007.0025
H-9985.0005BAEA 365.06 CAD
H-9985.0005BA H-9985.0025BA
Sex Pheromone Inhibitor iPD1 Trifluoroacetate
Proteins and Peptides
The octapeptide ALILTLVS is produced by Streptococcus faecalis donor strains harboring bacteriocin plasmid pPD1. It inhibits the activity of sex pheromone cPD1. The non-polar peptide iPD1 is also used for calibration in mass spectrometry.