Hi/Lo Profile Rocker, IBI Scientific
Supplier: IBI Scientific
ROCAA115S ROCLS0001 ROCLS0002 ROCLS0003
89408-326EA 2777.2 CAD
89408-326 89409-860 89409-862 89409-864
Hi/Lo Profile Rocker, IBI Scientific
Shakers Rotating/Rocking Shakers
The lightweight, low-profile rocker unit is designed for versatility
The low profile rocker fits easily into laboratory incubators, ovens, and refrigerated units without rearranging shelves or disturbing other materials. The thin, flat power cord reaches external outlets without damaging incubator or refrigerator gaskets. The platform is continuously adjustable from 2° to 11° and can be effortlessly changed in 15 seconds. The speed varies from 0 - 22rpm with no cogging at slow speeds.
All finishes and hardware are stainless steel and the rocker can be easily dissembled for sterilizingMore Product Information