SevenExcellence pH/ION Meters, METTLER TOLEDO®
Supplier: Mettler Toledo
30046248 30046249 30046250 30046251
CA89261-044EA 5048.18 CAD
CA89261-044 CA89261-046 CA89261-048 CA89261-050
SevenExcellence pH/ION Meters, METTLER TOLEDO®
Electrochemistry Meters pH and Multi-Parameter Meters
SevenExcellence pH/Ion meter redefines flexibility and offers various measurements such as pH, mV, rel. mV, and ion concentration in the form of one meter.
  • Multiparameter measurements
  • Touchscreen menu
For incremental ion measurements the instrument guides you through the various steps and does all the calculations for you.
USB-stick, LabX direct software data export.
Starting an analysis, changing the setting and accessing the results is easily achieved with the cleverly designed touch screen menu.
Order Now
SPECIFICATIONS
Environmentally Preferable
pH range ‒2.000 to 20.000
pH resolution 0.001/0.01/0.1
pH accuracy ±0.002
mV range ‒2000.0 to 2000.0
mV resolution 0.1/1
mV accuracy ±0.1
Temperature range ‒30.0...+130.0 °C
Temperature resolution 0.1 °C
Temperature accuracy ±0.1 °C
Display TFT
Data logging capacity 20000 data points, 250 analyses
Interfaces RS232, USB-A, USB-B, LAN, Mini-LTW, Mini-DIN, BNC, Cinch/RCA