SevenExcellence Conductivity Meters, METTLER TOLEDO®
Supplier: Mettler Toledo
30046244 30046245 30046246
CA89261-038EA 5255 CAD
CA89261-038 CA89261-040 CA89261-042
SevenExcellence Conductivity Meters, METTLER TOLEDO®
Electrochemistry Meters Conductivity Meters
SevenExcellence stands for convenient, easy-to-understand operation combined with high measurement accuracy and outstanding reliability.
  • Convenient operation
  • High measurement accuracy
  • Outstanding reliability
SevenExcellence can cope with complex applications and stringent regulations in regulated markets, but also feels at ease with routine measurements in the laboratory.
USB-stick, LabX direct software data export.
For special applications such as USP/EP and conductivity ash it guides you through the measurement steps and notifies you when you are out of range. Starting an analysis, changing the settings and accessing the results is easily achieved thanks to the cleverly designed touch screen menu.