FerroVer® Iron Reagent Powder Pillows, 5 ml, Hach
Supplier: Hach
92799
92746
CA927-99PK
64.54
CAD
CA927-99
CA11016-764
FerroVer® Iron Reagent Powder Pillows, 5 ml, Hach
Environmental Test Kits
For Iron determination by the 1,10 phenanthroline method, using FerroVer® Iron Reagent Powder Pillows.
For 5 ml samples.