BOD Seed Inoculum, POLYSEED, Hach
Supplier: Hach
2918700 2918700
CA24712-00PK 739.99 CAD
97026-776 CA24712-00
BOD Seed Inoculum, POLYSEED, Hach
Standards Biological Standards
BOD Seed Inoculum for use in Biochemical Oxygen Demand (BOD5) test procedures. A blend of specialized microbial cultures in an easy-to-use capsule. Designed to provide a uniform standard for the degradation of both industrial and municipal waste in BOD5 analyses.
  • Consitent seed source: same bacteria, same type of bacteria, same amount of bacteria blend
  • Known bacterial content is free from pathogens
  • Save and convenient pre-measured dry product in easy to use capsules
Polyseed®-NX CBOD contains an additive for the inhibition of ammonia nitrogen in a wastewater sample. Additive amount is comparable to that stated in Standard Methods as necessary for sufficient inhibition.
Two years shelf life
Application: Dilution water preparation. Method number: 8043
Ordering information: Package of 50 capsules.