SpecCheck Secondary Gel Standards Sets, DPD Chlorine, LR
Supplier: Hach
2635300
2635300
CA26353-00EA
569.24
CAD
CA26353-00
MSPP-2635300
SpecCheck Secondary Gel Standards Sets, DPD Chlorine, LR
Standards
Environmental Analysis Standards
DPD chlorine, low range, 0 to 2 mg/L as Cl₂.
- Fast and convenient
- Stable
Used to confirm consistent instrument response. Set contains four vials filled with gels that simulate the test color at various concentrations.
Ordering information: Set of four jars.