Bare Nichrome Wire
Supplier: Wards
PH1064D PH1064C PH1064B PH1064A
470149-224EA 67.75 CAD
470149-224 470149-226 470149-228 470149-230
Bare Nichrome Wire
Wire
Create High Performance Loops
125gm reels of bare Nichrome wire for use in resistance heating or inoculating loops.