Test Tube Basket
Supplier: Wards
CH0707A CH0707B
470137-792EA 39.7 CAD
470137-792 470137-794
Test Tube Basket
Baskets Baskets for Test Tubes
Safe Storage for Test Tubes of All Sizes
This test tube basket is made of polypropylene and is virtually unbreakable. You can use it with corrosive substances and easily autoclave the basket as well. Makes handling and storage of different sized test tubes a breeze. Size of small basket: 14 cm x 12 cm x 11 cm; Size of large basket: 16 cm x 16 cm x 16 cm.