Lattice Rod, Aluminum
Supplier: Wards
CH0658PLM3 CH0658PLM6
470140-274EA 9.49 CAD
470140-274 470140-276
Lattice Rod, Aluminum
Retort Rods
Resists corrosion
  • Made of T6 Temper 6063 aluminum alloy
These anodized, smooth, unthreaded aluminum rods are used for constructing lattice rod assemblies.