Distillation Apparatus, Short Path, Ace Glass
Supplier: Ace Glass
9316-02 9316-20 9316-04 9316-06 9316-08
89074-348EA 675.55 CAD
89074-348 89074-352 89074-356 89074-358 89074-360
Distillation Apparatus, Short Path, Ace Glass
Distillation Components Distillation Sets
A still designed for small quantities where holdup between flask and receiver must be minimized, but where it is also desired to measure vapor temperature with an accuracy compared to large apparatus
The upper hose connections are for water lines; the lower connection is for vacuum or vent. The center joint of the two-neck pear-shape flask is equipped with an adjustable gas inlet tube. All joints are [ST] 14/20. Hose connections are for use with 5/16-inch I.D. tubing.
Physical carry-over is practically eliminated by impingement. Condenser delivery tube is straight through. The condensate drops directly into one of the four receiver flasks attached to the "cow" receiver via the included Delrin clips. The "cow" receiver itself is held in place in the same way. Optional graduated receivers may be substituted for the pear shaped flasks.
Ordering information: Complete apparatus includes 89074-348 still body; 89074-356 "cow" receiver; 89074-358 gas inlet tube; four 5 mL 89074-912 receiver flasks; and 10 mL 89074-908 distillation flask. Optional parts include 89074-360 graduated receivers (four will be required) and 89075-032 10 mL heating mantle.