Still, Short Path, Minimum Hold-Up, Ace Glass
Supplier: Ace Glass
9315-08 9315-05 9315-10
89074-350EA 31.13 CAD
89074-350 89074-354 89074-370
Still, Short Path, Minimum Hold-Up, Ace Glass
Distillation Components Distillation Sets
This short path, minimum hold-up still is supplied with a one-piece, 89074-366 "cow" receiver with four 3 mL arms
Also available is an optional 89074-356 cow receiver that accommodates four 89074-912, 5mL mini-flasks. Standard taper joint sizes are 14/20, with a spherical 18/10 joint on the tube adapter and flask side neck. Gas inlet tube is positioned in the distillation flask with a 89051-100 Ace-Thred adapter. Use with a 5/16-inch I.D. tubing. Fabricated from borosilicate glass.
Ordering information: Complete apparatus includes 89074-354 still body; 89074-366 "cow" receiver; 10 mL 89More Product Information