SP Bel-Art ProCulture Warming Plate 37 °C, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F37015-0000 F37015-1000
30621-302EA 1086.63 CAD
30621-302 10126-432
SP Bel-Art ProCulture Warming Plate 37 °C, Bel-Art Products, a part of SP
Heaters Hot Plates
Increase cell viability during cell passaging with this highly accurate warming plate that keeps cells near an optimal 37°C (±1 °C) outside of the incubator.
  • Reduces cell stress and allows enzymes like trypsin to work efficiently
  • Can be used to maintain the warmth of reagents inside the hood
  • Fits easily in cell culture hoods or on the workbench
  • Temperature remains uniform across the entire anodized aluminum warming plate
  • 15-minute count down timer keeps track of time sensitive applications
  • UV resistant polypropylene housing will not break down under germicidal UV lights
  • Water resistant, safe for cleaning with ethanol or IPA
Avoids numerous entries into the CO2 incubator which alter its atmosphere, harming ongoing experiments in the incubated cultures.
Certifications: 220 V is CE marked.