Sublimation Apparatus, Ace Glass Incorporated
Supplier: Ace Glass
8022-06 8022-08 8022-10
89061-626EA 653.04 CAD
89061-626 89061-628 89061-630
Sublimation Apparatus, Ace Glass Incorporated
Condensers
A 50mm size for samples of approximately 25mL
With a #50 o-ring joint, the unit measures 200mm high x 50mm in diameter. The stopcock is positioned so that the bottom can be firmly clamped to the top. Fabricated from borosilicate glass.
Ordering information: Complete item includes outer body and inner top. These items may also be purchased separately. Use clamp 89060-266 (NOT included). For replacement o-ring, see 89061-038.