Classic Wood ADA Height Base Cabinets, Cupboard and Drawer Cabinets, Diversified Woodcrafts
Supplier: VWR International
89372-796 89372-814
89372-796EA 5589.34 CAD
89372-796 89372-814
Classic Wood ADA Height Base Cabinets, Cupboard and Drawer Cabinets, Diversified Woodcrafts
Furniture Casework
Classic oak laboratory grade cabinets.
  • Classic neutral color
Vertical grain matched select oak veneers finished with a VOC-free, environmentally friendly chemical-resistant finish. Cabinets stained with a light neutral finish. Plywood core cabinet body with removable backs, full extension drawer slides and stainless steel hinges. 96mm classic wire pulls, twin-pin seismic rated shelf pins, and nylon roller door catches.
Certifications: SEFA Tested.
Ordering information: Units are shipped wrapped and palletized via common carrier trucking. All cabinets include a Lifetime Warranty on manufacturing parts and processes.