Stopcock, Straight Bore, 1:5 PTFE Plug, Ace Glass Incorporated
Supplier: Ace Glass
8138-04 8138-06 8138-08 8138-10 8138-12
89062-334EA 151.76 CAD
89062-334 89062-336 89062-338 89062-340 89062-342
Stopcock, Straight Bore, 1:5 PTFE Plug, Ace Glass Incorporated
Stopcocks
A ruggedly constructed stopcock with a 1:5 PTFE plug and tapered flange, which ensures maximum strength at the seal
Bore on the side arms is equal to or slightly larger than the bores of the plug on all sizes. All units feature polished barrels. Fabricated from borosilicate glass.