Rodaviss® O-Rings, Ace Glass Incorporated
Supplier: Ace Glass
7617-02 7617-04 7617-06 7617-08 7617-10 7617-12 7617-13 7617-15 7617-17 7617-19 7617-21 7617-23
89059-726PK 34.4 CAD
89059-726 89059-728 89059-730 89059-732 89059-734 89059-736 89059-738 89059-740 89059-742 89059-744 89059-746 89059-748
Rodaviss® O-Rings, Ace Glass Incorporated
Seals Sealing Rings
Designed for use with Rodaviss® joints and caps to form a leak-tight seal
Available in Nitrile or Viton formats. For threaded joints, see 89059-696 series. For caps, see 89059-718 series.