KONTES® Nuts and Ferrules, KIMBLE®, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
420821-0018 420821-0116 420821-0618 420821-3519 420821-6116 420822-0018 420822-1019 420822-2007 420826-0018 420826-0116 420827-0018 420827-0116 420821-6218 420822-0116
KT420821-0018EA 15.23 CAD
KT420821-0018 KT420821-0116 KT420821-0618 KT420821-3519 KT420821-6116 KT420822-0018 KT420822-1019 KT420822-2007 KT420826-0018 KT420826-0116 KT420827-0018 KT420827-0116 KT420821-6218 KT420822-0116
KONTES® Nuts and Ferrules, KIMBLE®, DWK Life Sciences
Fittings and Connectors
Tubing nuts and ferrules are required to make the initial tubing connections to the end fittings of chromaflex AQ and chromaflex or columns.