Gas Dispersion Tube with Bottom Frit, Ace Glass Incorporated
Supplier: Ace Glass
7201-06 7201-12 7201-14 7201-16 7201-04 7201-02
89058-308EA 140 CAD
89058-308 89058-310 89058-312 89058-402 89058-404 89058-406
Gas Dispersion Tube with Bottom Frit, Ace Glass Incorporated
Gas Tubes
The outside diameter of this gas dispersion tube is 8mm, and the total length of the tube is 150mm
The bottom frit is approximately 2mm thick. Available in two different disc diameters, and three different porosities. Fabricated from borosilicate glass.