Arrow Air Stirrers, Medium and Heavy Duty, Ace Glass
Supplier: Ace Glass
13365-10 13365-15 13365-20
89077-662EA 1932 CAD
89077-662 76107-084 76107-086
Arrow Air Stirrers, Medium and Heavy Duty, Ace Glass
Stirrers Overhead Stirrers
Explosion proof air motors eliminate the possibility of spark ignition when mixing volatile chemicals, solvents, and other flammable liquids.
  • Explosion proof
  • Available in medium or heavy duty
  • Motors are equipped with a needle valve to control your mixing speed
  • Unit is complete and ready to mount on a laboratory stand
  • The muffler on the motor provides quiet operation
  • Stainless steel shaft and propeller resist most acids and chemicals, and allows for easier cleaning
  • Service costs are virtually eliminated, as the unit adjusts itself to compensate for wear
The air motors also provide increased speed and power for use with more viscous materials. A more constant speed is attained through gear reduction, lowering speed fluctuations due to changes in air pressure.