Straight Gas Dispersion Tube, Ace Glass Incorporated
Supplier: Ace Glass
7197-08 7197-02 7197-04 7197-06 7197-12 7197-14 7197-16 7197-18
89058-314EA 204.6 CAD
89058-314 89058-330 89058-332 89058-334 89058-348 89058-350 89058-352 89058-408
Straight Gas Dispersion Tube, Ace Glass Incorporated
Gas Tubes
The top tube features an 8mm O.D., and the total length of the tube is 150mm
Thickness of the bottom is 14mm. The bottom O.D. matches the disc diameter, and the overall length is 164mm. Available in two different disc diameters, and four different porosities. Fabricated from borosilicate glass.