Gas Dispersion Tube, Ace Glass Incorporated
Supplier: Ace Glass
7196-16 7196-04 7196-08 7196-10 7196-14 7196-12 7196-06 7196-20 7196-02 7196-18
89058-392EA 182.22 CAD
89058-392 89058-292 89058-294 89058-464 89058-298 89058-400 89058-300 89058-302 89058-304 89058-306
Gas Dispersion Tube, Ace Glass Incorporated
Gas Tubes
This gas dispersion tube's outside diameter is 8mm, and the total length of tube is 150mm
The bottom support is the same O.D. as the disc. Available in two different disc diameters, and five different porosities. Fabricated from borosilicate glass.