Straight Tube with Porus Disc, Ace Glass Incorporated
Supplier: Ace Glass
7205-12 7205-20 7205-26 7205-02 7205-04 7205-06 7205-08 7205-10 7205-14 7205-16 7205-30 7205-32 7205-34 7205-36 7205-38 7205-40 7205-44 7205-46 7205-52 7205-54 7205-56 7205-58 7205-72 7205-74 7205-76 7205-78 7205-22 7205-18 7205-24 7205-28 7205-42 7205-48
89058-288EA 119.48 CAD
89058-288 89058-296 89058-320 89058-382 89058-384 89058-386 89058-388 89058-390 89058-394 89058-396 89058-410 89058-412 89058-414 89058-416 89058-418 89058-420 89058-424 89058-426 89058-430 89058-432 89058-434 89058-436 89058-438 89058-440 89058-442 89058-444 89058-446 89058-466 89058-468 89058-470 89058-472 89058-474
Straight Tube with Porus Disc, Ace Glass Incorporated
Filters Filter Tubes
This tube is designed for the construction of special apparatus.
Disc is centered in tube. Available in various disc diameters and tube lengths. Frits available in five different porosities. Fabricated from borosilicate glass.