AccuStart™ II PCR SuperMix, QuantaBio

Supplier: Quantabio

95137-04K 95137-500 95137-100
CA89235-016EA 3715.07 CAD
CA89235-016 CA89235-018 CA76047-072
AccuStart™ II PCR SuperMix, QuantaBio
Nucleic Acid Reagents End-point PCR Enzymes and Kits

AccuStart II PCR SuperMix is a 2X concentrated, ready-to-use reaction cocktail for routine PCR amplification of DNA fragments up to 4 kb. It contains all components, except primers and template.


  • High yield, high sensitivity
  • Precise amplification - hot-start technology ensures specific and efficient primer extension
  • Superior reproducibility and precision
  • Convenient reaction assembly at room temperature


AccuStart II PCR SuperMix simplifies reaction assembly, improves assay reproducibility, and reduces the risk of contamination. A key component is AccuStart™ II Taq DNA polymerase which contains monoclonal antibodies that bind to the polymerase and keep it inactive prior to the initial PCR denaturation step. Upon heat activation (1 minute at 94°C), the antibodies denature irreversibly, releasing fully active, unmodified Taq DNA polymerase. This enables specific and efficient primer extension with the convenience of room temperature reaction assembly.


Contents:
- 2X reaction buffer containing 3 mM MgCl2
- 4 mM each dNTP (dATP, dCTP, dGTP, dTTP)
- AccuStart II Taq DNA Polymerase and stabilizers.


Storage conditions:
AccuStart II PCR SuperMix is stable for 1 year when stored in a constant temperature freezer at -20°C. For convenience, it may be stored unfrozen at +2 to +8°C for up to 12 months. Repeated freezing and thawing does not impair product performance.


Precautions and Disclaimer:
For research use only. Not intended for any animal or human therapeutic or diagnostic use. Please consult the Material Safety Data Sheet for information regarding hazards and safe handling practices.

Order Now

SPECIFICATIONS

Environmentally Preferable
Application

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR