AccuStart™ II GelTrack PCR SuperMix, QuantaBio
Supplier: Quantabio
95136-04K 95136-500 95136-100 95136-100
CA89235-012EA 2288.38 CAD
CA89235-012 CA89235-014 76047-140 CA76047-140
AccuStart™ II GelTrack PCR SuperMix, QuantaBio
Nucleic Acid Reagents End-point PCR Enzymes and Kits
AccuStart II GelTrack PCR SuperMix is a 2X concentrated, ready-to-use reaction cocktail for routine PCR amplification of DNA fragments up to 4 kb. It contains premixed loading dye to streamline gel electrophoresis and all components, except primers and template.
  • High yield; high sensitivity
  • Precise amplification
  • Superior reproducibility and precision
  • Convenient reaction assembly at room temperature
AccuStart II PCR SuperMix simplifies reaction assembly, improves assay reproducibility, and reduces the risk of contamination. A key component is AccuStart™ II Taq DNA polymerase which contains monoclonal antibodies that bind to the polymerase and keep it inactive prior to the initial PCR denaturation step. Upon heat activation (1 minute at 94 °C), the antibodies denature irreversibly, releasing fully active, unmodified Taq DNA polymerase. This enables specific and efficient primer extension with the convenience of room temperature reaction assembly.
Store components in a constant temperature freezer at −25 to −15 °C upon receipt. Repeated freezing and thawing does not impair product performance. For lot specific expiry date, refer to package label, Certificate of Analysis or Product Specification Form.
AccuStart II GelTrack PCR SuperMix is a 2X concentrated, ready-to-use reaction cocktail for routine PCR amplification of DNA fragments up to 4 kb. It contains premixed loading dye to streamline gel electrophoresis and all components, except primers and template.
Ordering information: AccuStart™ II GelTrack PCR Supermix contains 2X reaction buffer containing 3 mM MgCl₂, 0.4 mM each dNTP (dATP, dCTP, dGTP, dTTP), Accustart II Taq DNA Polymerase and GelTrack loading dye and stabilizers. For research only. Not intended for any animal or human therapeutic or diagnostic use. Please consult the Material Safety Data Sheet for information regarding hazards and safe handling practices.