Gasket for Ground Flanges, Ace Glass Incorporated
Supplier: Ace Glass
6495-10 6495-21 6495-23 6495-43 6495-47
89055-004EA 38.01 CAD
89055-004 89055-006 89055-008 89055-010 89055-012
Gasket for Ground Flanges, Ace Glass Incorporated
Seals Gaskets
Gaskets are designed for mating ground flange surfaces on reactor flasks and matching heads.
FETFE, an exclusive ACE product made from TFE impregnated fluoroelastomers, features a leak-free seal with clamp pressure and good chemical and temperature resistance. PTFE provides excellent chemical resistance and purity. The FETFE gaskets offer a thickness of 0.5 mm (0.02"), while the PTFE gaskets offer a thickness of 0.8 mm (0.03").
They can be used with all conical-style and flat flange-style flasks and heads. All gaskets feature grooves, with the exception of item 89055-004, which is designed without grooves. Available in either black FETFE or white PTFE.