Receivers, Moisture Test, Dean-Stark, Ace Glass
Supplier: Ace Glass
7720-08 7720-02 7720-04 7720-20
89060-328EA 503.48 CAD
89060-328 89060-338 89215-156 89060-332
Receivers, Moisture Test, Dean-Stark, Ace Glass
Distillation Components Distillation Receivers
This Dean-Stark moisture test receiver is suitable for ASTM D 95 and E 123
It is subdivided 0.1mL to 2mL and 0.2mL above 2mL. Fabricated from borosilicate glass.