Calpain inhibitor XI, solid

Supplier: Thermo Scientific Chemicals

J65991-LB0 J65991-MCR
CAAAJ65991-LB0EA 679.4 CAD
CAAAJ65991-LB0 CAAAJ65991-MCR
Calpain inhibitor XI, solid
Calpain inhibitor XI

Off-white solid


  • Light Sensitive


A cell-permeable dipeptidyl -ketoamide that acts as a potent, highly selective, reversible, and active site inhibitor of calpain-1 and -2

Formula: C₂₆H₄₀N₄O₆
Storage Temperature: Refrigerator
MDL Number: MFCD06411394
CAS Number: 145731-49-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR