Caspase-9 Inhibitor III

Supplier: Thermo Scientific
J65501-MCR J65501-MCR
CAAAJ65501-MCREA 590.92 CAD
AAJ65501-MCR CAAAJ65501-MCR
Caspase-9 Inhibitor III
Proteins and Peptides
CAS No.: 403848-57-7
Molecular Formula: C24H35ClN6O9
Formula Weight: 587.02
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White
MDL No.: MFCD02259690

A potent and irreversible inhibitor of caspase-9
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR