Compound E, solid

Supplier: Thermo Scientific Chemicals

Synonyms: (S)-2-(2-(3,5-Difluorophenyl)acetamido)-N-((S)-1-methyl-2-oxo-5-phenyl-2,3-dihydro-1H-benzo[e][1,4]diazepin-3-yl)propanamide

J65131-EXD J65131-EXD
CAAAJ65131-EXDEA 475.58 CAD
CAAAJ65131-EXD AAJ65131-EXD
Compound E, solid
Compound E

A cell-permeable, potent, selective, peptidomimetic, non-transition-state analog inhibitor of -secretase and Notch processing

Formula: C₂₇H₂₄F₂N₄O₃
MW: 490.511 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04974531
CAS Number: 209986-17-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR