Human Orexin B, Thermo Scientific
Supplier: Thermo Scientific Chemicals
J65110-EXD
J65110-MCR
CAAAJ65110-EXDEA
360.24
CAD
CAAAJ65110-EXD
CAAAJ65110-MCR
Human Orexin B, Thermo Scientific
Proteins and Peptides
Lyophilized powder
Molecular Formula: C123H212N44O35S
Formula Weight: 2899.33
Storage Temperature: -30°C to -10°C
Physical Form: Lyophilized powder
Appearance: White to off-white
Solubility: Soluble in water at 1mg/ml
MDL No.: MFCD01635373
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...