GAP27

Supplier: Thermo Scientific Chemicals

J64109-EXD J64109-MCR
CAAAJ64109-EXDEA 111.39 CAD
CAAAJ64109-EXD CAAAJ64109-MCR
GAP27
Proteins and Peptides

CAS No.: 198284-64-9
Molecular Formula: C60H101N15O17
Formula Weight: 1305.54
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Appearance: White to off-white
Solubility: Soluble at 1mg/ml in 1% acetic acid
MDL No.: MFCD03456920

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR