LB Agar Plate with Streptomycin, Thermo Scientific
Supplier: Thermo Scientific Chemicals
J62665-EQF
J63541-EQF
CAAAJ62665-EQFEA
339.31
CAD
CAAAJ62665-EQF
CAAAJ63541-EQF
LB Agar Plate with Streptomycin, Thermo Scientific
Media for Microbiology
Prepared Media for Microbiology
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...