Bombesin
Supplier: Thermo Scientific Chemicals
J63514-LB0
J63514-MCR
CAAAJ63514-LB0EA
582.15
CAD
CAAAJ63514-LB0
CAAAJ63514-MCR
Bombesin
Proteins and Peptides
CAS No.: 31362-50-2
Molecular Formula: C71H110N24O18S
Formula Weight: 1619.90
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
Solubility: Soluble in 0.05M acetic acid (20mg/ml)
Merck Reference: 14,1329
MDL No.: MFCD00167514
Gut tetradecapeptide with the ability to stimulate release of various hormones
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...