Midostaurin
Supplier: Thermo Scientific Chemicals
J62917-LB0
J62917-MCR
CAAAJ62917-LB0EA
1254.54
CAD
CAAAJ62917-LB0
CAAAJ62917-MCR
Midostaurin
Midostaurin
Inhibits a variety of serine/threonine and tyrosine kinases including PKC
CAS No.: 120685-11-2
Molecular Formula: C35H30N4O4
Formula Weight: 570.64
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White to off-white
Melting Point: 235-260°
Merck Reference: 14,6185
MDL No.: MFCD12828879
Inhibits a variety of serine/threonine and tyrosine kinases including PKC
Formula:
C₃₅H₃₀N₄O₄ Melting Pt: 235…260 °C Storage Temperature: Freezer |
MDL Number:
MFCD12828879 CAS Number: 120685-11-2 Merck Index: 14,6185 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...