Ionomycin 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
J62448-LB0
J62448-MCR
CAAAJ62448-LB0EA
779.19
CAD
CAAAJ62448-LB0
CAAAJ62448-MCR
Ionomycin 98%
Ionomycin
Solution in ethanol
Highly selective and potent calcium ionophore
Highly selective and potent calcium ionophore
Formula:
C₄₁H₇₂O₉ MW: 709.02 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD06798385 CAS Number: 56092-81-0 UN: 1993 ADR: 3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...