Magnesium sulfate 1 M in aqueous solution, sterile-filtered

Supplier: Thermo Scientific Chemicals

J61030-EQE J61030-AK
CAAAJ61030-EQEEA 150.07 CAD
CAAAJ61030-EQE CAAAJ61030-AK
Magnesium sulfate 1 M in aqueous solution, sterile-filtered
Magnesium sulphate

Liquid


CAS No.: 7487-88-9 Molecular Formula: MgO4S Formula Weight: 120.37 Physical Form: Liquid MDL No.: MFCD00011110

Formula: MgSO₄
Storage Temperature: Ambient
MDL Number: MFCD00011110
CAS Number: 7487-88-9
EINECS: 231-298-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1bc2ae189b0cbc1bfc599951528466503d0d9008&vid=63867221020ebd2c8c227492e609a4764639ff79&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR