12-Deoxyphorbol-13-acetate 99+%, powder

Supplier: Thermo Scientific Chemicals

J60971-LB0 J60971-MCR
CAAAJ60971-LB0EA 1469.42 CAD
CAAAJ60971-LB0 CAAAJ60971-MCR
12-Deoxyphorbol-13-acetate 99+%, powder
12-Deoxyphorbol-13-acetate

12-Deoxyphorbol 13-acetate; activator of protein kinase C. A weak mouse ear irritant, but lethal at 1ug per mouse


Purity: 99+% CAS No.: 60857-08-1 Molecular Formula: C22H30O6 Formula Weight: 390.47 Storage Temperature: -30°C to -10°C Physical Form: Powder Melting Point: 216-219° Solubility: Soluble in DMSO at 30mg/ml MDL No.: MFCD00674138


12-Deoxyphorbol 13-acetate; activator of protein kinase C. A weak mouse ear irritant, but lethal at 1ug per mouse

Formula: C₂₂H₃₀O₆
Melting Pt: 216…219 °C
Storage Temperature: Freezer
MDL Number: MFCD00674138
CAS Number: 60857-08-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR